Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271313.1 | NP_001258242.1 | 21 | 4308 | ||
XM_008770553.1 | XP_008768775.1 | 21 | 4370 | |||
XM_008770554.1 | XP_008768776.1 | 21 | 4370 | |||
GenBank mRNA | FQ214753.1 | - | 3 | 332 |