Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108385.1 | NP_001101855.1 | 10, 11 | 1176 | ||
XM_006252279.2 | XP_006252341.1 | 9, 10 | 1311 | |||
XM_006252280.2 | XP_006252342.1 | 9, 10 | 1309 | |||
XM_006252281.1 | XP_006252343.1 | 7, 8 | 664 | |||
XM_008770830.1 | XP_008769052.1 | 10, 11 | 1174 | |||
XM_008770831.1 | XP_008769053.1 | 10, 11 | 1831 | |||
GenBank mRNA | BC166453.1 | - | 9, 10 | 1185 | 8374835 |