Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108388.1 | NP_001101858.1 | 25 | 1591 | ||
XM_006252428.2 | XP_006252490.1 | 26 | 1673 | |||
XM_008770882.1 | XP_008769104.1 | 26 | 1655 | |||
XM_008770883.1 | XP_008769105.1 | 25 | 1616 |