Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108393.1 | NP_001101863.1 | 23 | 3526 | ||
XM_006252537.2 | XP_006252599.1 | 22 | 16037 | |||
XM_006252538.2 | XP_006252600.1 | 23 | 3573 |