Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Purg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191554.1 | NP_001178483.1 | 1 | 251 | ||
XM_006253233.2 | XP_006253295.1 | 1 | 249 | |||
XM_006253234.2 | XP_006253296.1 | 1 | 249 | |||
XM_006253235.2 | XP_006253297.1 | 1 | 249 |