Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044247.1 | NP_001037712.1 | 5 | 646 | ||
XM_006253100.2 | XP_006253162.1 | 5 | 2076 | |||
XM_008771246.1 | XP_008769468.1 | 5 | 1079 | |||
XM_008771247.1 | XP_008769469.1 | 5 | 674 | |||
XM_008771248.1 | XP_008769470.1 | 5 | 1919 | |||
GenBank mRNA | BC098935.1 | - | 5 | 646 | 7445963 | |
FQ215658.1 | - | 5 | 518 |