Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MPDZ SIRNA Assays ›
Gene Symbol:
MPDZ
Gene Name:
multiple PDZ domain crumbs cell polarity complex component
Gene Aliases:
HYC2, MUPP1
Chromosome Location:
Chr.9: 13105296 - 13279687 on Build GRCh38
UniGene ID:
Hs.169378
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16720
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MPDZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261406.1
NP_001248335.1
32
4610
NM_001261407.1
NP_001248336.1
32
4610
NM_003829.4
NP_003820.2
33
4709
XM_005251622.4
XP_005251679.1
33
4658
XM_005251623.4
XP_005251680.1
31
4448
XM_006716885.3
XP_006716948.1
33
4707
XM_006716886.3
XP_006716949.1
34
4866
XM_006716887.3
XP_006716950.1
33
4810
XM_006716888.3
XP_006716951.1
33
4658
XM_006716889.3
XP_006716952.1
32
4562
XM_006716891.3
XP_006716954.1
32
4562
XM_017015252.1
XP_016870741.1
33
4658
XM_017015253.1
XP_016870742.1
32
4547
XM_017015254.1
XP_016870743.1
32
4659
XM_017015255.1
XP_016870744.1
31
4507
XM_017015256.1
XP_016870745.1
31
4448
XM_017015257.1
XP_016870746.1
31
4435
GenBank mRNA
AB210041.1
-
33
4710
AF093419.1
-
32
4533
AK074721.1
-
3
194
AK091945.1
-
8
987
AK294823.1
-
6
619
AK294976.1
-
11
1215
AK307924.1
-
6
619
AK307988.1
-
9
1027
AK316489.1
-
10
1093
BC140793.1
-
31
4434
9021661
BC144564.1
-
31
4434
9053092
CR936648.1
-
17
4597
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
viral process
myelination
protein binding
protein C-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0