Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286485.1 | NP_001273414.1 | 16 | 1660 | ||
NM_001286486.1 | NP_001273415.1 | 15 | 1528 | |||
NM_003834.2 | NP_003825.1 | 17 | 1704 | |||
NM_183337.2 | NP_899180.1 | 17 | 1693 | |||
XM_011522719.2 | XP_011521021.1 | 18 | 1787 | |||
XM_011522720.2 | XP_011521022.1 | 18 | 1727 | |||
GenBank mRNA | AB209582.1 | - | 9 | 3600 | ||
AF035153.1 | - | 17 | 1664 | |||
AF035154.1 | - | 17 | 1675 | |||
AK091701.1 | - | 15 | 1525 | |||
BC141803.1 | - | 17 | 1668 | 40147243 |