Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748694.3 | XP_003748742.1 | 4 | 2497 | ||
XM_006227909.2 | XP_006227971.1 | 4 | 2497 | |||
XM_006227910.2 | XP_006227972.1 | 4 | 2497 | |||
XM_006227911.2 | XP_006227973.1 | 4 | 2497 | |||
XM_008758741.1 | XP_008756963.1 | 4 | 2497 | |||
XM_008758742.1 | XP_008756964.1 | 4 | 2497 | |||
GenBank mRNA | BC169129.1 | - | 2 | 247 |