Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isoc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008367.1 | NP_001008368.1 | 5 | 603 | ||
XM_006228274.2 | XP_006228336.1 | 6 | 857 | |||
GenBank mRNA | BC085336.1 | - | 5 | 603 | 7301753 |