Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199328.2 | NP_001186257.2 | 2 | 222 | ||
NM_001199329.2 | NP_001186258.2 | 2 | 222 | |||
NM_003838.4 | NP_003829.3 | 2 | 222 | |||
GenBank mRNA | AF017445.1 | - | 2 | 192 | ||
AK223254.1 | - | 2 | 189 | |||
AK294948.1 | - | 2 | 194 | |||
AK299468.1 | - | 2 | 167 | |||
AK304490.1 | - | 2 | 190 | |||
BC020720.1 | - | 2 | 196 | |||
BC032308.1 | - | 2 | 178 | 5184469 | ||
DB066016.1 | - | 2 | 226 | |||
KU178439.1 | - | 2 | 155 | |||
KU178440.1 | - | 2 | 155 | |||
KU178441.1 | - | 2 | 155 |
Gene Symbol: FPGT-TNNI3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112808.2 | NP_001106279.2 | 2 | 222 | ||
NM_001199327.1 | NP_001186256.2 | 2 | 222 | |||
GenBank mRNA | AB208941.1 | - | 2 | 171 | ||
BC032865.1 | - | 2 | 195 | 5263275 | ||
BX640903.1 | - | 2 | 207 |