Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108474.1 | NP_001101944.1 | 2 | 479 | ||
GenBank mRNA | BC099083.1 | - | 2 | 438 | ||
BC158846.1 | - | 2 | 464 | 7443540 |