Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108485.1 | NP_001101955.1 | 5 | 750 | ||
XM_006229235.1 | XP_006229297.1 | 5 | 1021 |