Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271573.1 | NP_001258502.1 | 2 | 285 | ||
NM_003847.2 | NP_003838.1 | 3 | 401 | |||
GenBank mRNA | AB015594.1 | - | 3 | 344 | ||
AF093668.1 | - | 3 | 306 | |||
AK001415.1 | - | 3 | 394 | |||
AK297442.1 | - | 2 | 133 | |||
AK313318.1 | - | 3 | 315 | |||
BC009697.1 | - | 3 | 295 | 3878406 | ||
BC101164.2 | - | 3 | 255 | 40015908 | ||
BC114384.1 | - | 3 | 255 | 40004153 | ||
CR542046.1 | - | 3 | 249 | |||
CR542075.1 | - | 3 | 249 | |||
DA773492.1 | - | 2 | 285 |