Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUCLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003850.2 | NP_003841.1 | 4 | 528 | ||
XM_017020806.1 | XP_016876295.1 | 4 | 748 | |||
XM_017020807.1 | XP_016876296.1 | 6 | 763 | |||
XM_017020808.1 | XP_016876297.1 | 4 | 515 | |||
XM_017020809.1 | XP_016876298.1 | 5 | 612 | |||
GenBank mRNA | AB035863.1 | - | 4 | 528 | ||
AF058953.1 | - | 3 | 360 | |||
AF087890.1 | - | 4 | 536 | |||
AK001458.1 | - | 4 | 426 | |||
AK001847.1 | - | 4 | 515 | |||
AK225452.1 | - | 4 | 515 | |||
AK299437.1 | - | 4 | 467 | |||
AK302311.1 | - | 5 | 587 | |||
AK315513.1 | - | 4 | 490 | |||
AK316271.1 | - | 5 | 843 | |||
BC027587.2 | - | 4 | 480 | 4829291 | ||
BX640602.1 | - | 4 | 499 | |||
KU178442.1 | - | 4 | 471 |