Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Snx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230940.2 | XP_006231002.1 | 2 | 323 | ||
XM_006230941.2 | XP_006231003.1 | 2 | 326 | |||
XM_008760201.1 | XP_008758423.1 | 1 | 707 | |||
XM_008760202.1 | XP_008758424.1 | 1 | 707 | |||
XM_341993.7 | XP_341994.5 | 2 | 321 | |||
GenBank mRNA | FQ222518.1 | - | 2 | 263 |