Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212529.1 | NP_997694.1 | 4 | 474 | ||
XM_006255964.2 | XP_006256026.1 | 3 | 552 | |||
XM_006255965.2 | XP_006256027.1 | 4 | 589 | |||
XM_006255966.2 | XP_006256028.1 | 3 | 552 | |||
GenBank mRNA | BC091424.1 | - | 2 | 696 |