Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lemd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039032.1 | NP_001034121.1 | 4 | 531 | ||
GenBank mRNA | BC098775.1 | - | 4 | 531 | 7373953 |
Gene Symbol: NEWGENE_1305341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772790.1 | XP_008771012.1 | 2 | 214 |