Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfesd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108540.1 | NP_001102010.1 | 3 | 285 | ||
XM_006231706.2 | XP_006231768.1 | 4 | 1385 | |||
XM_006231708.2 | XP_006231770.1 | 4 | 814 | |||
XM_006231709.2 | XP_006231771.1 | 4 | 499 |