Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxadrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199406.2 | NP_955438.1 | 5 | 741 | ||
XM_008766539.1 | XP_008764761.1 | 5 | 1528 | |||
XM_008766540.1 | XP_008764762.1 | 4 | 848 | |||
GenBank mRNA | BC063181.1 | - | 5 | 746 | 6887452 |
Gene Symbol: LOC103690070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766579.1 | XP_008764801.1 | 1528 | |||
XM_008766580.1 | XP_008764802.1 | 1528 | ||||
XM_008766581.1 | XP_008764803.1 | 848 |