Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760646.1 | XP_008758868.1 | 6, 7 | 605 | ||
XM_008760647.1 | XP_008758869.1 | 6, 7 | 605 | |||
XM_008760648.1 | XP_008758870.1 | 6, 7 | 604 | |||
XM_008760649.1 | XP_008758871.1 | 6, 7 | 604 | |||
XM_008760650.1 | XP_008758872.1 | 5, 6 | 515 | |||
XM_008760651.1 | XP_008758873.1 | 5, 6 | 514 | |||
XM_008760652.1 | XP_008758874.1 | 5, 6 | 514 | |||
XM_008760653.1 | XP_008758875.1 | 5, 6 | 513 | |||
GenBank mRNA | FQ221531.1 | - | 6, 7 | 504 |