Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101669.1 | NP_001095139.1 | 25 | 3053 | ||
NM_003866.2 | NP_003857.2 | 26 | 3152 | |||
XM_005263323.3 | XP_005263380.1 | 18 | 2261 | |||
XM_005263325.3 | XP_005263382.1 | 18 | 2234 | |||
XM_011532391.2 | XP_011530693.1 | 26 | 3164 | |||
XM_011532392.2 | XP_011530694.1 | 25 | 2903 | |||
XM_017008794.1 | XP_016864283.1 | 26 | 3164 | |||
XM_017008795.1 | XP_016864284.1 | 25 | 2960 | |||
XM_017008796.1 | XP_016864285.1 | 24 | 2933 | |||
XM_017008797.1 | XP_016864286.1 | 19 | 2284 | |||
XM_017008798.1 | XP_016864287.1 | 19 | 2261 | |||
GenBank mRNA | AK298118.1 | - | 18 | 2203 | ||
AK302936.1 | - | 25 | 2906 | |||
BC017924.2 | - | 5 | 539 | |||
BC110918.1 | - | 24 | 2910 | 6160633 | ||
BC133005.1 | - | 22 | 2608 | 40147027 | ||
BC144255.1 | - | 23 | 2703 | |||
BX649090.1 | - | 26 | 2993 | |||
U96922.1 | - | 22 | 2678 |