Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003876.2 | NP_003867.1 | 2 | 755 | ||
NR_024547.1 | 3 | 1047 | ||||
GenBank mRNA | AK311965.1 | - | 2 | 321 | ||
BC002819.2 | - | 2 | 343 | 3636199 | ||
BC005268.1 | - | 2 | 755 | 4051155 | ||
BC020815.1 | - | 2 | 321 | 4296149 | ||
BT006768.1 | - | 2 | 312 | |||
X51804.1 | - | 2 | 570 |
Gene Symbol: ENO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298600.1 | - | 613 |