Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108589.1 | NP_001102059.1 | 16 | 2033 | ||
XM_006234781.2 | XP_006234843.1 | 15 | 2390 |