Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277235.1 | NP_001264164.1 | 18, 19 | 2233 | ||
XM_006235126.2 | XP_006235188.1 | 19, 20 | 2426 | |||
XM_006235127.2 | XP_006235189.1 | 19, 20 | 2415 | |||
GenBank mRNA | FQ211753.1 | - | 18, 19 | 2233 | ||
FQ219827.1 | - | 4, 5 | 466 |