Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108594.1 | NP_001102064.1 | 3 | 268 | ||
XM_006235128.2 | XP_006235190.1 | 3 | 308 | |||
GenBank mRNA | BC159413.1 | - | 3 | 251 | 8367451 | |
FQ214547.1 | - | 3 | 240 |