Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135801.1 | NP_001129273.1 | 18 | 2094 | ||
GenBank mRNA | BC088338.1 | - | 6 | 609 | ||
BC160885.1 | - | 18 | 2094 | 7384442 |