Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047914.1 | NP_001041379.1 | 1, 2 | 134 | ||
GenBank mRNA | AY387078.1 | - | 1, 2 | 134 |