Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003884.4 | NP_003875.3 | 14 | 2534 | ||
XM_005265528.4 | XP_005265585.1 | 14 | 2159 | |||
XM_017007423.1 | XP_016862912.1 | 15 | 1989 | |||
XM_017007424.1 | XP_016862913.1 | 14 | 2085 | |||
GenBank mRNA | AL832173.1 | - | 4 | 447 | ||
BC060823.1 | - | 14 | 2139 | 30336561 | ||
BC070075.1 | - | 14 | 2204 | 30333414 | ||
U57317.2 | - | 14 | 2546 |