Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236925.2 | XP_006236987.1 | 12 | 3768 | ||
XM_006236926.2 | XP_006236988.1 | 12 | 3765 |