Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238141.2 | XP_006238203.1 | 4 | 1816 | ||
XM_006238142.2 | XP_006238204.1 | 3 | 985 | |||
XM_008763689.1 | XP_008761911.1 | 4 | 1645 | |||
XM_342829.7 | XP_342830.4 | 1595 |