Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033709.1 | NP_001028881.1 | 3 | 298 | ||
GenBank mRNA | BC101850.1 | - | 3 | 298 | 5621707 | |
FQ210318.1 | - | 3 | 313 |