Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749992.3 | XP_003750040.1 | 15 | 2250 | ||
XM_003749993.3 | XP_003750041.1 | 15 | 2250 | |||
XM_006238567.2 | XP_006238629.1 | 17 | 2415 | |||
XM_006238570.2 | XP_006238632.1 | 17 | 2415 | |||
XM_006238571.2 | XP_006238633.1 | 16 | 2373 | |||
XM_006238574.2 | XP_006238636.1 | 14 | 1869 | |||
XM_008763937.1 | XP_008762159.1 | 17 | 2412 | |||
XM_008763938.1 | XP_008762160.1 | 16 | 2376 | |||
XM_008763939.1 | XP_008762161.1 | 16 | 2373 | |||
XM_008763941.1 | XP_008762163.1 | 16 | 2292 | |||
XM_008763942.1 | XP_008762164.1 | 15 | 2253 | |||
XM_008763943.1 | XP_008762165.1 | 16 | 2034 | |||
XM_008763944.1 | XP_008762166.1 | 16 | 2031 | |||
XM_008763945.1 | XP_008762167.1 | 17 | 2489 | |||
XM_008763946.1 | XP_008762168.1 | 15 | 1908 | |||
XM_008763948.1 | XP_008762170.1 | 14 | 1869 | |||
XM_342877.7 | XP_342878.5 | 15 | 1995 |