Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134956.1 | NP_001128428.1 | 5 | 2080 | ||
XM_008764151.1 | XP_008762373.1 | 3 | 1806 | |||
XM_008764152.1 | XP_008762374.1 | 3 | 1701 | |||
XM_008764153.1 | XP_008762375.1 | 3 | 1844 | |||
XM_008764154.1 | XP_008762376.1 | 3 | 1820 |