Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173432.1 | NP_001166903.1 | 14 | 1429 | ||
XM_006239246.2 | XP_006239308.1 | 14 | 1502 | |||
XM_006239247.2 | XP_006239309.1 | 14 | 1499 | |||
XM_006239248.2 | XP_006239310.1 | 14 | 1502 | |||
XM_006239249.2 | XP_006239311.1 | 14 | 2130 | |||
XM_006239250.2 | XP_006239312.1 | 14 | 1502 | |||
XM_006239251.2 | XP_006239313.1 | 14 | 1499 | |||
XM_006239252.2 | XP_006239314.1 | 14 | 1502 | |||
GenBank mRNA | BC168672.1 | - | 14 | 1429 | 7935928 | |
FQ212004.1 | - | 14 | 1468 |