Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108737.1 | NP_001102207.1 | 11, 12 | 1130 | ||
XM_006240977.2 | XP_006241039.1 | 11, 12 | 1365 | |||
XM_006240978.1 | XP_006241040.1 | 11, 12 | 1152 |