Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241463.2 | XP_006241525.1 | 6 | 1190 | ||
XM_008765404.1 | XP_008763626.1 | 5 | 881 | |||
XM_008765405.1 | XP_008763627.1 | 6 | 1185 | |||
XM_008765406.1 | XP_008763628.1 | 6 | 1020 | |||
XM_008765407.1 | XP_008763629.1 | 5 | 881 |