Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173558.1 | NP_001167029.1 | 3 | 356 | ||
XM_006241605.2 | XP_006241667.1 | 3 | 315 | |||
GenBank mRNA | EV763930.1 | - | 3 | 356 | ||
FQ228874.1 | - | 3 | 366 | |||
FQ229670.1 | - | 1 | 15 |