Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173558.1 | NP_001167029.1 | 6, 7 | 777 | ||
XM_006241605.2 | XP_006241667.1 | 6, 7 | 757 | |||
GenBank mRNA | BC158730.1 | - | 4, 5 | 371 | ||
FQ228874.1 | - | 6, 7 | 787 | |||
FQ229670.1 | - | 4, 5 | 436 |