Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108748.1 | NP_001102218.1 | 2 | 786 | ||
XM_006242217.2 | XP_006242279.1 | 2 | 1192 |