Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035000.1 | NP_001030172.1 | 3, 4 | 525 | ||
XM_006242218.1 | XP_006242280.1 | 3, 4 | 1434 | |||
XM_006242219.1 | XP_006242281.1 | 3, 4 | 1434 | |||
XM_006242220.1 | XP_006242282.1 | 3, 4 | 1434 | |||
GenBank mRNA | BC092573.1 | - | 3, 4 | 525 | 7323327 |