Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079855.1 | NP_001073324.1 | 6 | 840 | ||
NM_001184702.1 | NP_001171631.1 | 6 | 840 | |||
NM_001184703.1 | NP_001171632.1 | 7 | 933 | |||
NM_001184704.1 | NP_001171633.1 | 5 | 698 | |||
NM_003918.2 | NP_003909.2 | 7 | 933 | |||
XM_011545600.2 | XP_011543902.1 | 6 | 771 | |||
XM_017029927.1 | XP_016885416.1 | 7 | 883 | |||
XM_017029928.1 | XP_016885417.1 | 7 | 858 | |||
XM_017029929.1 | XP_016885418.1 | 7 | 1043 | |||
XM_017029930.1 | XP_016885419.1 | 6 | 748 | |||
XM_017029931.1 | XP_016885420.1 | 6 | 771 | |||
GenBank mRNA | AK098046.1 | - | 5 | 606 | ||
AK292496.1 | - | 6 | 748 | |||
BC023152.1 | - | 6 | 707 | 3923041 | ||
DQ424901.1 | - | 5 | 651 | |||
U94358.1 | - | 3 | 329 | |||
U94360.1 | - | 3 | 388 | |||
U94361.1 | - | 2 | 106 | |||
U94362.1 | - | 7 | 934 | |||
U94363.1 | - | 6 | 714 |