Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012202.2 | NP_001012202.2 | 2 | 2488 | ||
XM_006243633.2 | XP_006243695.1 | 3 | 2790 | |||
XM_006243634.2 | XP_006243696.1 | 2 | 1214 | |||
GenBank mRNA | BC087097.1 | - | 2 | 563 |