Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012202.2 | NP_001012202.2 | 10 | 3285 | ||
XM_006243633.2 | XP_006243695.1 | 11 | 3587 | |||
XM_006243634.2 | XP_006243696.1 | 10 | 2011 | |||
GenBank mRNA | BC087097.1 | - | 10 | 1360 |