Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750579.2 | XP_003750627.1 | 4 | 462 | ||
XM_008766592.1 | XP_008764814.1 | 3 | 226 | |||
XM_008766593.1 | XP_008764815.1 | 4 | 462 |