Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008859.1 | NP_001008859.1 | 3, 4 | 218 | ||
XM_006244464.2 | XP_006244526.1 | 4, 5 | 367 | |||
XM_006244465.2 | XP_006244527.1 | 4, 5 | 364 | |||
GenBank mRNA | AY310148.1 | - | 3, 4 | 218 |