Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750650.3 | XP_003750698.1 | 22, 23 | 2368 | ||
XM_006244582.2 | XP_006244644.1 | 21, 22 | 2330 | |||
XM_006244583.1 | XP_006244645.1 | 21, 22 | 2410 | |||
XM_008766903.1 | XP_008765125.1 | 22, 23 | 2358 |