Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271050.1 | NP_001257979.1 | 1 | 1189 | ||
XM_006244827.2 | XP_006244889.1 | 1 | 1135 |