Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276270.1 | NP_001263199.1 | 7 | 1957 | ||
NM_001276271.1 | NP_001263200.1 | 7 | 1975 | |||
NM_001276273.1 | NP_001263202.1 | 6 | 1021 | |||
NM_003925.2 | NP_003916.1 | 7 | 1975 | |||
XM_011513268.2 | XP_011511570.1 | 6 | 769 | |||
GenBank mRNA | AF072250.1 | - | 7 | 1805 | ||
AF114784.1 | - | 7 | 1770 | |||
AK303013.1 | - | 7 | 1831 | |||
AK312878.1 | - | 7 | 1713 | |||
AM180876.1 | - | 6 | 675 | |||
BC011752.2 | - | 7 | 1654 | 3534047 | ||
BC034463.1 | - | 7 | 1629 | |||
CD678412.1 | - | 3 | 234 | |||
CR450305.1 | - | 7 | 1611 |