Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100865.1 | NP_001094335.1 | 2 | 218 | ||
GenBank mRNA | BC097401.1 | - | 2 | 218 |